Cole-Parmer Tube Sinker, PVDF (Kynar), 3/4" Hose Barb
Representative image only.
Cole-Parmer Tube Sinker, PVDF (Kynar), 3/4" Hose Barb
Cole-Parmer | Item: 5014294
Weighted sinkers help keep tubing at the bottom of container
₹19,400.00INR / Each
₹19,400.00INR / Each
Product Description
Key Features
  • Flow-through design—perfect for filling and dispensing
  • PDVF (Kynar) construction offers superior chemical resistance
  • Hose barb fitting accepts flexible tubing
  • ASME-BPE, USP Class VI, FDA 21 CFR 177.2510, REACH, and RoHS compliant
  • Animal-derived ingredient free (ADIF) and animal-derived component free (ADCF)
  • Sterilize by autoclave, gamma irradiation, x-ray, or EtO
  • 3.12"H x 1.13" dia
  • 3/4" hose barb
More About this Item
Connect a tube sinker to your tubing to keep it at the bottom of your container. The flow-through design provides efficient emptying or filling by pumps or dispensers. Slight recess at the bottom of the sinker prevents it from suctioning against the container. Monobarb fitting accepts flexible tubing.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Sterilization MethodAutoclave, gamma irradiation, x-ray, or EtO
  • Connection TypeHose Barb
  • Barb Size / Tubing Id (Port 1) (in)3/4
  • ColorTranslucent
  • Min Temperature (° F)-80
  • Max Temperature (° F)275
  • Min Temperature (° C)-62
  • Max Temperature (° C)135
  • Regulatory ApprovalsNon-Phthalate;RoHS;ADCF;FDA;USP
  • DescriptionTube Sinker, PVDF (Kynar), 3/4" Hose Barb