Cole-Parmer® Essentials Transfer Pipettes, Individual Wrap, Sterile
No more breakage or cross-contamination
  • Translucent shatterproof LDPE is nontoxic and biologically inert
  • Nonbinding for cells and proteins to prevent sample loss
  • Heat sealable and liquid nitrogen compatible
Low-density polyethylene (LDPE) makes these sterile pipettes with built-in bulbs the perfect substitute for breakable glass pipettes, eliminating cross-contamination and cumbersome rubber bulb changes. Plastic reduces the loss of cells and proteins that often occurs with binding that takes place with glass alternatives. Precision manufacturing ensures uniform repeatable drops.
Heat seal pipettes to provide liquid-tight transport and storage. These pipettes are suitable for use within a temperature range of -58 to 194°F (-50 to 90°C). Come sterile and individually wrapped.
11,500.00 - ₹15,800.00INR / Box of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Max Volume (mL) Overall Length (in)
Availability Pricing
COMPARE
LDPE 3 5.45
₹11,500.00INR / Box of 500
COMPARE
LDPE 5 6.125
₹11,500.00INR / Box of 500
COMPARE
LDPE 6 9.1
₹15,800.00INR / Box of 500
COMPARE
LDPE 7.5 6.125
₹12,900.00INR / Box of 500