Cole-Parmer® Threaded to Hose Barb Fitting, Tee Adapter, PP
Threaded barb adapter design provides safe, difficult-to-break connection.
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Easily combine or split fluid flow using tee unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. However, it’s important to avoid using them with oxidizing agents, as these can compromise their integrity.
1,800.00 - ₹3,100.00INR / Pkg of 10
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
COMPARE
1/8 1/8 1/8
₹3,100.00INR / Pkg of 10
COMPARE
1/4 1/4 1/8
₹1,800.00INR / Pkg of 10
COMPARE
3/8 3/8 1/8
₹1,800.00INR / Pkg of 10
COMPARE
1/4 1/4 1/4
₹1,800.00INR / Pkg of 10
COMPARE
3/8 3/8 1/4
₹1,800.00INR / Pkg of 10
COMPARE
1/2 1/2 1/4
₹1,900.00INR / Pkg of 10
COMPARE
1/4 1/4 3/8
₹1,900.00INR / Pkg of 10
COMPARE
3/8 3/8 3/8
₹1,900.00INR / Pkg of 10
COMPARE
1/2 1/2 3/8
₹1,900.00INR / Pkg of 10
COMPARE
1/4 1/4 1/2
₹2,000.00INR / Pkg of 10
COMPARE
3/8 3/8 1/2
₹2,000.00INR / Pkg of 10
COMPARE
1/2 1/2 1/2
₹2,000.00INR / Pkg of 10
COMPARE
3/4 3/4 1/2
₹2,400.00INR / Pkg of 10
COMPARE
1/4 1/4 3/4
₹2,400.00INR / Pkg of 10
COMPARE
3/8 3/8 3/4
₹2,400.00INR / Pkg of 10
COMPARE
1/2 1/2 3/4
₹2,400.00INR / Pkg of 10
COMPARE
3/4 3/4 3/4
₹2,400.00INR / Pkg of 10
COMPARE
1 1 3/4
₹2,700.00INR / Pkg of 10
COMPARE
1 1 1
₹2,700.00INR / Pkg of 10
Showing 1- 19 of 19