Cole-Parmer Threaded to Hose Barb Fitting, Tee Adapter, PP, 1'' NPT(M) x 1" ID; 10/Pk
Representative image only.
Cole-Parmer Threaded to Hose Barb Fitting, Tee Adapter, PP, 1'' NPT(M) x 1" ID; 10/Pk
Cole-Parmer | Item: 5974226
Threaded barb adapter design provides safe, difficult-to-break connection.
₹2,700.00INR / Pkg of 10
₹2,700.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Easily combine or split fluid flow using tee unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. However, it’s important to avoid using them with oxidizing agents, as these can compromise their integrity.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeNPT(M) x Hose Barb
  • Number Of Ports3
  • Thread Size NPT(M) (Port 1) (InchShort)1
  • Barb Size / Tubing Id (Port 2) (in)1
  • Barb Size / Tubing Id (Port 3) (in)1
  • Regulatory ApprovalsFDA;RoHS
  • DescriptionThreaded to Hose Barb Fitting, Tee Adapter, PP, 1" NPT(M) x 1" ID; 10/Pk