Cole-Parmer® Threaded to Hose Barb Fitting, Straight Adapter, Natural PVDF
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks. These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
1,500.00 - ₹2,900.00INR / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Thread size UNF(M) (port 1) (InchShort)
Availability Pricing
COMPARE
1/8 PVDF 1/4"-28
₹2,200.00INR / Pkg of 10
COMPARE
5/32 PVDF 1/4"-28
₹2,600.00INR / Pkg of 10
COMPARE
3/16 PVDF 1/4"-28
₹2,900.00INR / Pkg of 10
COMPARE
1/16 PVDF 10-32
₹1,500.00INR / Pkg of 10
COMPARE
3/32 PVDF 10-32
₹1,600.00INR / Pkg of 10