Cole-Parmer Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar, 10-32 UNF(M) x 3/32" ID; 10/Pk
Representative image only.
Cole-Parmer Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar, 10-32 UNF(M) x 3/32" ID; 10/Pk
Cole-Parmer | Item: 5974207
Threaded barb adapter design provides safe, difficult-to-break connection
₹1,600.00INR / Pkg of 10
₹1,600.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks. These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Sterilization MethodAutoclave
  • Number Of Ports2
  • Connection TypeUNF (M) x Hose Barb
  • Thread size UNF(M) (port 1) (InchShort)10-32
  • Barb Size / Tubing Id (Port 2) (in)3/32
  • Flow PatternStraight
  • ColorNatural
  • Regulatory ApprovalsRoHS
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, Natural Kynar, 10-32 UNF(M) x 3/32" ID; 10/Pk