Cole-Parmer® Threaded Tee Adapter, Natural Kynar
Discover durable, PFAS-free solutions with this versatile Kynar conversion nipple for precise fluid transfer and secure connections
  • Made from high-quality, durable Kynar material for chemical resistance
  • PFAS-free, ensuring environmental safety and compliance
  • Compact yet sturdy design with a lightweight build
  • Ideal for industrial, lab, and medical applications requiring reliable fluid connections
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Easily combine or split fluid flow using tee unions.  These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability. Natural Kynar®, also known as natural Polyvinylidene Fluoride (PVDF), is a PFAS-free resin.
5,000.00 - ₹12,500.00INR / Pkg of 10
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
COMPARE
1/8 1/8 1/8
₹5,700.00INR / Pkg of 10
COMPARE
1/4 1/4 1/8
₹5,000.00INR / Pkg of 10
COMPARE
3/8 3/8 1/8
₹5,200.00INR / Pkg of 10
COMPARE
1/4 1/4 1/4
₹5,600.00INR / Pkg of 10
COMPARE
3/8 3/8 1/4
₹5,700.00INR / Pkg of 10
COMPARE
1/2 1/2 1/4
₹6,000.00INR / Pkg of 10
COMPARE
1/4 1/4 3/8
₹6,200.00INR / Pkg of 10
COMPARE
3/8 3/8 3/8
₹6,400.00INR / Pkg of 10
COMPARE
1/2 1/2 3/8
₹6,600.00INR / Pkg of 10
COMPARE
1/4 1/4 1/2
₹6,800.00INR / Pkg of 10
COMPARE
3/8 3/8 1/2
₹7,000.00INR / Pkg of 10
COMPARE
1/2 1/2 1/2
₹7,300.00INR / Pkg of 10
COMPARE
3/4 3/4 1/2
₹7,700.00INR / Pkg of 10
COMPARE
1/4 1/4 3/4
₹7,900.00INR / Pkg of 10
COMPARE
3/8 3/8 3/4
₹8,200.00INR / Pkg of 10
COMPARE
1/2 1/2 3/4
₹8,500.00INR / Pkg of 10
COMPARE
3/4 3/4 3/4
₹9,600.00INR / Pkg of 10
COMPARE
1 1 3/4
₹11,700.00INR / Pkg of 10
COMPARE
1 1 1
₹12,500.00INR / Pkg of 10
Showing 1- 19 of 19