Cole-Parmer Threaded to Hose Barb Fitting, Tee Adapter, Natural Kynar, 1/2" NPT(M) x 1/2" ID; 10/Pk
Representative image only.
Cole-Parmer Threaded to Hose Barb Fitting, Tee Adapter, Natural Kynar, 1/2" NPT(M) x 1/2" ID; 10/Pk
Cole-Parmer | Item: 5974276
Discover durable, PFAS-free solutions with this versatile Kynar conversion nipple for precise fluid transfer and secure connections
₹7,300.00INR / Pkg of 10
₹7,300.00INR / Pkg of 10
Product Description
Key Features
  • Made from high-quality, durable Kynar material for chemical resistance
  • PFAS-free, ensuring environmental safety and compliance
  • Compact yet sturdy design with a lightweight build
  • Ideal for industrial, lab, and medical applications requiring reliable fluid connections
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Easily combine or split fluid flow using tee unions.  These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability. Natural Kynar®, also known as natural Polyvinylidene Fluoride (PVDF), is a PFAS-free resin.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Sterilization MethodAutoclave
  • Connection TypeNPT(M) x Hose Barb
  • Number Of Ports3
  • Thread Size NPT(M) (Port 1) (InchShort)1/2
  • Barb Size / Tubing Id (Port 2) (in)1/2
  • Barb Size / Tubing Id (Port 3) (in)1/2
  • Flow PatternTee
  • ColorNatural
  • Max Temperature (° F)285
  • Max Temperature (° C)140
  • Regulatory ApprovalsUSP;RoHS;ADCF
  • DescriptionThreaded to Hose Barb Fitting, Tee Adapter, Natural Kynar, 1/2" NPT(M) x 1/2" ID; 10/Pk