Cole-Parmer® ThinTouchPROtect™ Nitrile Gloves
Advanced Composition with Higher Breakthrough Time
  • Tested for use with 29 chemotherapeutic drugs per the ASTM D6978-05 Available in three different sizes: 9.5", 12" & 16" Enhanced chemical resistance Exceptional breakthrough time: Carmustine (3.3 mg/ml): 127.5 mins ThioTepa (10.0 mg/ml): 240 mins 0.65 AQL Level 3 Static dissipative with resistivity of 1x10^7 Ohms at humidity 51% Higher tensile strength for puncture and tear resistance Textured fingertip for exceptional wet and dry grip
Stay protected without compromising on comfort!
Use Cole-Parmer® ThinTouchPROtect™ Nitrile Gloves for worry free protection against cytotoxic chemicals, microbes and viruses.
11,900.00 - ₹18,200.00INR / Case of 500
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size Description
Availability Pricing
COMPARE
12 Small Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 12" (S) 500/Cs
₹11,900.00INR / Case of 500
COMPARE
12 Medium Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 12" (M) 500/Cs
₹11,900.00INR / Case of 500
COMPARE
12 Large Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 12" (L) 500/Cs
₹11,900.00INR / Case of 500
COMPARE
12 X-Large Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 12" (XL) 500/Cs
₹11,900.00INR / Case of 500
COMPARE
9 1/2 Small Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 9.5" (S), 1000/Cs
₹14,400.00INR / Case of 1000
COMPARE
9 1/2 Medium Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 9.5" (M), 1000/Cs
₹14,400.00INR / Case of 1000
COMPARE
9 1/2 Large Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 9.5" (L), 1000/Cs
₹14,400.00INR / Case of 1000
COMPARE
9 1/2 X-Large Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 9.5" (XL), 1000/Cs
₹14,400.00INR / Case of 1000
COMPARE
16 Small Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 16" (S), 500/Cs
₹18,200.00INR / Case of 500
COMPARE
16 Medium Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 16" (M), 500/Cs
₹18,200.00INR / Case of 500
COMPARE
16 Large Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 16" (L), 500/Cs
₹18,200.00INR / Case of 500
COMPARE
16 X-Large Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 16" (XL), 500/Cs
₹18,200.00INR / Case of 500
Showing 1- 12 of 12