Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 9.5" (XL); 1000/CS

Representative image only.

Cole-Parmer ThinTouchPROtect™ Nitrile Gloves, 9.5" (XL); 1000/CS

Cole-Parmer | Item: 1597226

Advanced Composition with Higher Breakthrough Time

₹14,400.00INR / Case of 1000

₹14,400.00INR / Case of 1000
Product Description

Key Features

  • Tested for use with 29 chemotherapeutic drugs per the ASTM D6978-05 Available in three different sizes: 9.5", 12" & 16" Enhanced chemical resistance Exceptional breakthrough time: Carmustine (3.3 mg/ml): 127.5 mins ThioTepa (10.0 mg/ml): 240 mins 0.65 AQL Level 3 Static dissipative with resistivity of 1x10^7 Ohms at humidity 51% Higher tensile strength for puncture and tear resistance Textured fingertip for exceptional wet and dry grip

More About this Item

Stay protected without compromising on comfort!
Use Cole-Parmer® ThinTouchPROtect™ Nitrile Gloves for worry free protection against cytotoxic chemicals, microbes and viruses.

Conformite Europeenne
Specifications
  • MaterialNitrile
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • DescriptionCole-Parmer ThinTouchPROtect™ Nitrile Gloves, 9.5" (XL), 1000/Cs
Reviews
Q&A