Cole-Parmer® Testing Sieves, 12" dia, Stainless Steel Frame/Stainless Steel Wire
Get more accurate results
  • Sievers are serialized
  • Come with a test sieve certificate of compliance
These stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.
42,800.00 - ₹98,900.00INR / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (InchShort) Sieve Opening (mm) U.S. Standard Screen Size (No.)
Availability Pricing
COMPARE
3.25 2.36 8
₹47,000.00INR / Each
COMPARE
3.25 2 10
₹47,000.00INR / Each
COMPARE
3.25 1.68 12
₹47,000.00INR / Each
COMPARE
3.25 1.41 14
₹47,000.00INR / Each
COMPARE
3.25 1.19 16
₹47,000.00INR / Each
COMPARE
3.25 1 18
₹42,800.00INR / Each
COMPARE
3.25 0.841 20
₹47,000.00INR / Each
COMPARE
3.25 0.707 25
₹47,000.00INR / Each
COMPARE
3.25 0.595 30
₹47,000.00INR / Each
COMPARE
3.25 0.5 35
₹47,000.00INR / Each
COMPARE
3.25 0.425 40
₹47,000.00INR / Each
COMPARE
3.25 0.355 45
₹47,000.00INR / Each
COMPARE
3.25 0.297 50
₹47,000.00INR / Each
COMPARE
3.25 0.25 60
₹47,000.00INR / Each
COMPARE
3.25 0.212 70
₹47,000.00INR / Each
COMPARE
3.25 0.177 80
₹47,000.00INR / Each
COMPARE
3.25 0.149 100
₹47,000.00INR / Each
COMPARE
3.25 0.125 120
₹47,000.00INR / Each
COMPARE
3.25 0.105 140
₹50,300.00INR / Each
COMPARE
3.25 0.088 170
₹53,000.00INR / Each
Showing 1 - 20 of 50