Cole-Parmer® Submersible 3 Foot ORP Electrode
Reach further into solutions
  • Water/wastewater, plating baths
  • Three-foot length for easier measuring in tanks
  • Sturdy design—permanently potted in sturdy ABS plastic housing
  • Submersible up to cable
Submersible electrodes operate from 32 to 176°>F (0 to 80°>C). Submerse up to the cable. Electrodes are potted in a 3 ft L x 1-1/16'' diameter ABS sheath. These electrodes are ideal for measurements in open containers such as tanks or troughs.
32,000.00 - ₹33,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Basic ph/mv/temperature benchtop meter, 110 vac
₹33,600.00INR / Each
COMPARE
Submersible 3' L ORP Probe, DJ/ABS/10'/BNC
₹32,000.00INR / Each