Cole-Parmer® Sterile Serological Pipettes, Polystyrene (PS), Individually Wrapped
Achieve quick and accurate fluid dispensing for all applications
  • Each pipette has color-coded “to deliver“ (TD) rings for easy identification
  • Bi-directional graduations on the pipettes provide added applicability
  • Graduations show volume removed or remaining, on all pipettes larger than 1ml
  • Gamma sterilized
  • Designed for quick and accurate fluid dispensing
These serological pipettes are made from transparent bio-compatible polystyrene (PS). Unique filter plug eliminates lint fibres and ensures the plug stays in place to prevent overfilling. Each lot is tested for endotoxins and cytotoxicity, and guaranteed non-haemolytic—ideal for tissue culture applications. The highly legible permanent graduations prevent ambiguity and guarantee precise dispensing to ±2%. Generous negative graduations provide additional pipetting volume.
The individually wrapped pipettes are supplied in a fiber-free paper/plastic combination that is easy to open. The dispenser box allows you to remove pipettes without opening the top. Universal stacking design requires less laboratory space and each box is color-coded for quick recognition.
5,300.00 - ₹13,300.00INR / Case of 200
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Increments (mL) Max Volume (mL)
Availability Pricing
COMPARE
0.01 1
₹8,800.00INR / Case of 500
COMPARE
0.1 1
₹9,700.00INR / Case of 500
COMPARE
0.01 2
₹13,300.00INR / Case of 500
COMPARE
0.1 5
₹5,300.00INR / Case of 200
COMPARE
0.1 10
₹6,600.00INR / Case of 200
COMPARE
0.2 25
₹7,800.00INR / Case of 150
COMPARE
0.5 50
₹12,400.00INR / Case of 100
Showing 1- 7 of 7