Cole-Parmer® Sterile Latex Gloves G Series
Minimize Risk and Maximize Productivity with Cole-Parmer Sterile Latex Gloves
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Gamma sterilised to ensure best-in-class performance Optimized for pharmaceutical manufacturing Pair packed non particulate-shedding pouch Ultra low-protein content AQL* 1.5
Cole-Parmer® latex gloves are ergonomically designed and manufactured from quality natural rubber latex for superior tactile sensitivity and comfort. They are powder-free and treated for low-protein to minimise allergic reactions. Unlike
commercially available medical gloves – these gloves are optimised for pharmaceutical manufacturing. Gloves feature fully textured fi nish that enhance operator grip – even when the gloves are wet. This is especially useful in pharmaceutical
manufacturing where the operators are required to repeatedly wet the gloves with IPA. Gloves have beaded cuff for ease of donning. Low-particulate last-mile packaging is used to avoid challenging cleanroom integrity. Last mile packaging
(pair pouches) carry comprehensive product information, part number identifi cation, size and lot number details to avoid operator-errors in glove selection.
8,300.00 - ₹11,400.00INR / Case of 600
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
COMPARE
12 Small
₹8,300.00INR / Case of 600
COMPARE
12 Medium
₹8,300.00INR / Case of 600
COMPARE
12 Large
₹8,300.00INR / Case of 600
COMPARE
12 X-Large
₹8,300.00INR / Case of 600
COMPARE
14 Small
₹10,800.00INR / Case of 600
COMPARE
14 Medium
₹11,400.00INR / Case of 600
COMPARE
14 Large
₹11,400.00INR / Case of 600
COMPARE
14 X-Large
₹11,400.00INR / Case of 600
COMPARE
16 Small
₹8,800.00INR / Case of 400
COMPARE
16 Medium
₹8,800.00INR / Case of 400
COMPARE
16 Large
₹8,800.00INR / Case of 400
COMPARE
16 X-Large
₹8,800.00INR / Case of 400
COMPARE
18 Small
₹10,400.00INR / Case of 400
COMPARE
18 Medium
₹10,400.00INR / Case of 400
COMPARE
18 Large
₹10,400.00INR / Case of 400
COMPARE
18 X-Large
₹10,400.00INR / Case of 400
Showing 1- 16 of 16