Cole-Parmer® Sterile Latex Gloves E-Series
Minimize Risk and Maximize Productivity with Cole-Parmer Sterile Latex Gloves
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Optimized for pharmaceutical manufacturing Manufactured under strict GMP Pair packed non particulate-shedding pouch Low-protein latex gloves AQL* 1.5 Available in four different sizes: 12, 14, 16 and 18 inch
Cole-Parmer® latex gloves are ergonomically designed and manufactured from quality natural rubber latex for superior tactile sensitivity and comfort. They are powder-free and treated for low-protein to minimise allergic reactions. Unlike commercially available medical gloves – these gloves are optimised for pharmaceutical manufacturing. Gloves feature fully textured fi nish that enhance operator grip – even when the gloves are wet. This is especially useful in pharmaceutical manufacturing where the operators are required to repeatedly wet the gloves with IPA. Gloves have beaded cuff for ease of donning. Low-particulate last-mile packaging is used to avoid challenging cleanroom integrity. Last mile packaging
(pair pouches) carry comprehensive product information, part number identifi cation, size and lot number details to avoid operator-errors in glove selection
8,900.00 - ₹21,700.00INR / Case of 1200
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size Description
Availability Pricing
COMPARE
12 Small Cole-Parmer E-Series 12" S Latex Gloves, 600 pairs
₹16,600.00INR / Case of 1200
COMPARE
12 Medium Cole-Parmer E-Series 12" M Latex Gloves, 600 pairs
₹16,600.00INR / Case of 1200
COMPARE
12 Large Cole-Parmer E-Series 12" L Latex Gloves, 600 pairs
₹16,600.00INR / Case of 1200
COMPARE
12 X-Large Cole-Parmer E-Series 12" XL Latex Gloves, 600 pairs
₹16,600.00INR / Case of 1200
COMPARE
16 Small Cole-Parmer E-Series 16" S Latex Gloves, 200 pairs
₹8,900.00INR / Case of 400
COMPARE
16 Medium Cole-Parmer E-Series 16" M Latex Gloves, 200 pairs
₹8,900.00INR / Case of 400
COMPARE
16 Large Cole-Parmer E-Series 16" L Latex Gloves, 200 pairs
₹8,900.00INR / Case of 400
COMPARE
16 X-Large Cole-Parmer E-Series 16" XL Latex Gloves, 200 pairs
₹8,900.00INR / Case of 400
COMPARE
14 Small Cole-Parmer E-Series 14" S Latex Gloves, 600 pairs
₹21,700.00INR / Case of 1200
COMPARE
14 Medium Cole-Parmer E-Series 14" M Latex Gloves, 600 pairs
₹21,700.00INR / Case of 1200
COMPARE
14 Large Cole-Parmer E-Series 14" L Latex Gloves, 600 pairs
₹21,700.00INR / Case of 1200
COMPARE
14 X-Large Cole-Parmer E-Series 14" XL Latex Gloves, 600 pairs
₹21,700.00INR / Case of 1200
COMPARE
18 Small Cole-Parmer E-Series 18" S Latex Gloves, 200 pairs
₹10,600.00INR / Case of 400
COMPARE
18 Medium Cole-Parmer E-Series 18" M Latex Gloves, 200 pairs
₹10,600.00INR / Case of 400
COMPARE
18 Large Cole-Parmer E-Series 18" L Latex Gloves, 200 pairs
₹10,600.00INR / Case of 400
COMPARE
18 X-Large Cole-Parmer E-Series 18" XL Latex Gloves, 200 pairs
₹10,600.00INR / Case of 400
Showing 1- 16 of 16