Cole-Parmer® Standard Laboratory Support Stand with Rod
Standard support base and rod assemblies are made entirely of 303 stainless steel. Stands consist of a sturdy base that can accommodate vessels up to 18” (45.7 cm) in diameter within the “U” shaped base and one removable rod. The base is also designed to hold up to three rods; a center rod and one rod per leg.
97,700.00 - ₹315,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accepts Rod Diameter (in) Length (in) Description
Availability Pricing
COMPARE
0.625 28 Support Stand Stainless steel with 28” Rod
₹97,700.00INR / Each
COMPARE
0.625 23 Support Stand Stainless Steel with 23” Rod
₹126,000.00INR / Each
COMPARE
0.625 36 Support Stand Stainless Steel with 36” Rod
₹146,000.00INR / Each
COMPARE
0.625 40 Support Stand Stainless Steel with 40” Rod
₹205,000.00INR / Each
COMPARE
0.625 48 Support Stand Stainless Steel with 48” Rod
₹287,000.00INR / Each
COMPARE
0.625 60 Support Stand Stainless Steel with 60” Rod
₹315,000.00INR / Each