Cole-Parmer® Standard Heavy-Duty Support Stand with Rod
Heavy-Duty support base and rod assemblies are made with a cast iron base and a 303 stainless steel rod. The stands base is designed to accommodate vessels up to 12” (30.5 cm) in diameter so to sit close to the support rod. Each of the corners of the four legs base rests on a rubber cushion for added stability. Stand features a corrosion-resistant, electro polished cast iron base.
61,000.00 - ₹145,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accepts Rod Diameter (in) Length (in) Rod Material
Availability Pricing
COMPARE
0.625 18 Stainless Steel
₹61,000.00INR / Each
COMPARE
0.625 23 Stainless Steel
₹113,000.00INR / Each
COMPARE
0.625 28 Stainless Steel
₹72,200.00INR / Each
COMPARE
0.625 36 Stainless Steel
₹77,400.00INR / Each
COMPARE
0.625 40 Stainless Steel
₹145,000.00INR / Each
COMPARE
0.625 48 Stainless Steel
₹86,800.00INR / Each
COMPARE
0.625 60 Stainless Steel
₹95,000.00INR / Each
Showing 1- 7 of 7