Cole-Parmer® Stainless Steel Crucibles with Covers
No welded seams, no weaknesses
  • Constructed from a solid piece of 304 stainless steel
  • Withstands temperatures from –292 to 1472°F (–180 to 800°C)
  • Cover is included
Unibody construction from Type 304 stainless steel. Can withstand temperatures from -180°C to 800°C. Covers included.
2,600.00 - ₹5,300.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (mm) Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
COMPARE
34 15 32
₹2,600.00INR / Each
COMPARE
34 20 33
₹2,700.00INR / Each
COMPARE
34 25 36
₹2,700.00INR / Each
COMPARE
38 30 39
₹2,900.00INR / Each
COMPARE
55 50 46
₹3,900.00INR / Each
COMPARE
55 100 57
₹5,300.00INR / Each