Cole-Parmer® Stackable Graduated Polypropylene Beakers
Won’t stick together
  • Stacking ribs prevent sticking for easier grabbing Autoclavable
Stackable polypropylene (PP) beakers are graduated in both metric and English (cc and oz.) units. Beakers are flexible — a gently squeeze creates a spout anywhere on the rim. Beakers can withstand temperatures from 0 to 121°C (32 to 250°F).
4,300.00 - ₹92,300.00INR / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
120 10 Polypropylene(PP)
₹4,300.00INR / Pkg of 25
COMPARE
500 50 Polypropylene(PP)
₹8,500.00INR / Pkg of 25
COMPARE
120 10 Polypropylene(PP)
₹87,700.00INR / Pkg of 1000
COMPARE
500 50 Polypropylene(PP)
₹92,300.00INR / Pkg of 500