Cole-Parmer® StableTemp™ Spark Free Refrigerators
Designed for safe storage of samples with explosion proof interior with grounded metal parts
  • Explosion-proof interior with EU explosion proof ATEX II C-T6
  • Setting temperature range +3°C to +16°C
  • Micro-processor control system with LED display
  • Multiple alarms and data function
  • Energy efficient design
  • Warranty: 1 year on main unit, 3 years on compressor
Cole-Parmer StableTemp Spark Free refrigerators are ideal for storing samples at below ambient condition in explosion-proof interior. Models are with forced air cooling with Auto defrost system along with multiple alarms and data function enabling you to check and save historical data. The carbon-hydrogen cooling system doubles the cooling efficiency and saves half the energy when compared to traditional refrigerants.
191,000.00 - ₹534,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (Hz) Power (VAC) Description
Availability Pricing
COMPARE
50 240 Spark free Refrigerator 10.9 CuFt Antistatic; 240 VAC, 50 Hz; 1/Ea
₹534,000.00INR / Each
COMPARE
50 240 Spark Free Refrigerator, 4.2 CuFt, 220-240 VAC, 50 Hz; 1/Ea
₹191,000.00INR / Each
COMPARE
50 240 Spark Free Refrigerator, 4.2 CuFt, Antistatic, 220-240 VAC, 50 Hz; 1/Ea
₹463,000.00INR / Each
COMPARE
50 240 Spark Free Refrigerator, 10.9 CuFt, 220-240 VAC, 50 Hz; 1/Ea
₹271,000.00INR / Each