Cole-Parmer® StableTemp™Combination Refrigerator/Freezer
Excellent performance with fast cooling speed
  • USB interface for data download
  • Independent displays for refrigerator and freezer sections
  • Multiple alarms to detect malfunctions
  • Compressor warranty: 3 years
Designed for optimal performance, this upright unit is capable of recording temperature data for up to one month automatically. Optional remote alarm and monitor via Internet.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Freezer Capacity (cu ft) Refrigerator Capacity (cu ft)
Availability Pricing
COMPARE
230 3.43 6.53
Call For Price
COMPARE
230 3.43 6.53