Cole-Parmer® StableTemp™ -30°C Low-Temperature Freezers
LED digital display for easy observation
  • Forced-air cooling with internal fan for maximum temperature uniformity and stability
  • Cooling fan is permanently lubricated for long life
  • Automatic defrost maintains optimum cooling capacity (model 16340-10)
  • Compressor warranty: 3 years
Upright freezers are scientifically designed to minimize moisture accumulation. Controller with digital display is conveniently mounted in the front panel and recessed to prevent an accidental setting change. Multiple alarm system for safe and reliable operation and alerts temperature rise or power loss. Freezers have built-in microprocessor controlled PT100 sensor with remote alarm port. Superior refrigeration system for pull-down time from 25°C ambient to –30°C in 170 minutes and warm-up time from 25°C ambient –30°C to –20°C is 160 minutes.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Power (Hz) Power (VAC)
Availability Pricing
COMPARE
10 50 230
Call For Price
COMPARE
12.3 50 230
Call For Price
COMPARE
17.3 50 230
Call For Price
COMPARE
14.8 50 230