Cole-Parmer® Solid Laboratory Black Rubber Stoppers
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.
9,200.00 - ₹48,900.00INR / Pkg of 181
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Standard Size Number
Availability Pricing
COMPARE
000
₹12,500.00INR / Pkg of 181
COMPARE
0
₹10,900.00INR / Pkg of 77
COMPARE
1
₹9,200.00INR / Pkg of 57
COMPARE
2
₹9,500.00INR / Pkg of 52
COMPARE
3
₹10,200.00INR / Pkg of 39
COMPARE
4
₹10,800.00INR / Pkg of 33
COMPARE
5
₹12,400.00INR / Pkg of 28
COMPARE
5.5
₹12,300.00INR / Pkg of 26
COMPARE
6.5
₹12,800.00INR / Pkg of 18
COMPARE
7
₹10,500.00INR / Pkg of 15
COMPARE
7.5
₹12,900.00INR / Pkg of 14
COMPARE
8
₹9,800.00INR / Pkg of 12
COMPARE
9
₹12,000.00INR / Pkg of 10
COMPARE
9.5
₹11,700.00INR / Pkg of 9
COMPARE
10
₹12,200.00INR / Pkg of 8
COMPARE
10.5
₹12,000.00INR / Pkg of 7
COMPARE
11
₹12,800.00INR / Pkg of 6
COMPARE
11.5
₹14,100.00INR / Pkg of 5
COMPARE
12
₹10,600.00INR / Pkg of 5
COMPARE
13
₹10,800.00INR / Pkg of 4
Showing 1 - 20 of 25