Cole-Parmer Soil Analysis Hydrometers
  • NIST Traceable
  • Temperature of Standardization 68°F/68°F
  • Conform to ASTM Specifications
  • Metal Ballast
  • Individually Serialized
Ideal for determining the specific gravity of soil, meets ASTM specifications. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
9,400.00 - ₹10,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0.995/1.038 Soil Analysis ASTM 151H Hydrometer
₹10,600.00INR / Each
COMPARE
-5/60g/L Soil Analysis ASTM 152H Hydrometer
₹9,400.00INR / Each