Cole-Parmer® SHP-200 Series Large-Capacity Digital Stirring Hot Plates
Strong stirring and precise heating for your large-volume needs
  • Powerful magnets for strong magnetic coupling
  • Microprocessor controlled, dual thermocouples ensure accurate temperature control
  • Large plate area accommodates one vessel up to 15 L
  • Easy-to-use temperature and stirring control dials
  • Hot warning light remains active even when switched off, but still connected to the mains supply
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
These stirring hot plates are designed with safety as well as performance in mind. Powerful magnets and motor give stirring speeds up to 2000 rpm and can mix large volumes up to 15 L*. The electronic feedback speed control accurately maintains the set speed. All models include an integral fitting for attaching a support rod.
Microprocessor control allows for accurate plate temperature monitoring. The "Hot" warning light will flash whenever the plate temperature is above 50 °C. An independent safety circuit protects against overheating and internal electronic components are protected against corrosion.
The housing is shaped for stability deflecting spills away from the user. Units also feature BioCote antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.
138,000.00 - ₹149,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Material Top Plate Width (in)
Availability Pricing
COMPARE
230 Glass Ceramic 12
₹143,000.00INR / Each
COMPARE
120 Glass Ceramic 12
₹149,000.00INR / Each
COMPARE
230 Al/Si Metal Alloy 12
₹138,000.00INR / Each
COMPARE
120 Al/Si Metal Alloy 12
₹146,000.00INR / Each