Cole-Parmer® Sealing Mats for Deep-Well Plates
Prevent spillage and contamination between wells
  • Tight seal reduces evaporation losses
  • Easily pierceable with pipette tips
  • Free of RNase, DNase, and endotoxins
  • Suitable for 96-well deep well plates
Choose from TPE or EVA materials to meet your application needs.
48,500.00 - ₹88,700.00INR / Pkg of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
COMPARE
TPE Sealing Mat for 96-Well Plates, 0.5- and 1.2 mL; 50/pk
₹88,700.00INR / Pkg of 50
COMPARE
EVA Sealing Mat for 96-Well Plates, 2.2 mL; 50/pk
₹48,500.00INR / Pkg of 50