Cole-Parmer Sealing Mat for 96-Well Plates, 2.2 mL, EVA; 50/pk
Representative image only.
Cole-Parmer Sealing Mat for 96-Well Plates, 2.2 mL, EVA; 50/pk
Cole-Parmer | Item: 6710470
Prevent spillage and contamination between wells
₹48,500.00INR / Pkg of 50
₹48,500.00INR / Pkg of 50
Product Description
Key Features
  • Tight seal reduces evaporation losses
  • Easily pierceable with pipette tips
  • Free of RNase, DNase, and endotoxins
  • Suitable for 96-well deep well plates
More About this Item
Choose from TPE or EVA materials to meet your application needs.
Specifications
  • MaterialEVA
  • DescriptionSealing Mat for 96-Well Plates, 2.2 mL; 50/pk