Cole-Parmer® Sanitizing Floor Mats
Decontaminate footwear before and after entry to designated areas, providing critical protection against contamination
  • Rubber fingers scrub the crevices in soles of shoes
  • Disinfectant solution sanitizes footwear surfaces
  • Chemically resistant to common disinfectant solutions
  • Suggested Disinfectant : Commonly used solutions include quaternary disinfectants or a diluted chlorine solution
  • Part no. 15978-36 holds 1 gallon of disinfectant
  • Part no. 15978-37 holds 5 gallons of disinfectant
Cleaning and Maintenance:
High-pressure hose (not to exceed 500 psi) and hot water (max 160o F) to rid the mats of oils. For best results use a mild soap or detergent with a pH between 4 and 9, to clean the mats. Do not use steam (unless specifically recommended), degreasers, or caustic chemicals. Do not machine wash or mechanically scrub the mats.
4,300.00 - ₹6,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Width (in) Description
Availability Pricing
COMPARE
32 24 Sanitizing Floor Mat, 24" x 32", Black; 1/Ea
₹4,300.00INR / Each
COMPARE
39 32 Sanitizing Floor Mat, 32" x 39"; Black; 1/Ea
₹6,000.00INR / Each