Cole-Parmer® Sanitary Gaskets, Black Buna
Compatible with Tri-Clamp® style fittings
  • Excellent chemical resistance—suitable for CIP
  • Resistant to many hydrocarbons, fats, and greases
  • Usable temperature range: –34 to 93 °C (–30 to 200 °F)
  • Phthalate free
  • FDA CFR 21 177 2600 and 3A compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Buna, a synthetic rubber compound, has excellent resistance to many hydrocarbons, fats, oils, greases, hydraulic fluids, and chemicals. Use between temperatures ranging from –34 to 93 °C (–30 to 200 °F). Gaskets are phthalate free and withstand long-term exposure to CIP media.
1,500.00 - ₹2,600.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sanitary Gasket Size (in) Seal Material Description
Availability Pricing
COMPARE
1/2 Buna Sanitary Gasket, Black Buna, 1/2"; 10/Pk
₹2,300.00INR / Pkg of 10
COMPARE
3/4 Buna Sanitary Gasket, Black Buna, 3/4"; 10/Pk
₹2,600.00INR / Pkg of 10
COMPARE
1 Buna Sanitary Gasket, Black Buna, 1"; 10/Pk
₹1,700.00INR / Pkg of 10
COMPARE
1 1/2 Buna Sanitary Gasket, Black Buna, 1-1/2"; 10/Pk
₹1,600.00INR / Pkg of 10
COMPARE
2 Buna Sanitary Gasket, Black Buna, 2"; 10/Pk
₹1,500.00INR / Pkg of 10
COMPARE
2 1/2 Buna Sanitary Gasket, Black Buna, 2-1/2"; 10/Pk
₹1,800.00INR / Pkg of 10