Cole-Parmer® Rotary Evaporator
Quality, efficient evaporator at an affordable price
  • Digital control for reproducible results Versatile—can be orientated for left or right handedness Simple, counterbalanced lift mechanism PTFE/glass liquid pathway for chemical inertness Long-life graphite impregnated PTFE vacuum seal Efficient flask and vapour tube ejection system
Our Cole-Parmer® evaporator range offers simple control and three glassware configurations—diagonal, vertical, or cold finger condensing depending on your application and space requirements. All configurations are available with plastic-coated glass* for additional safety.
Choose the diagonal condenser where height space is limited, while the vertical condenser is suitable where bench space is limited or in fume hoods. The cold finger condenser is the option to select if you don’t have a constant water supply for cooling the evaporated solvent.
Different Types:
Glass set 00
Diagonal coil condenser, vapour tube for diagonal condenser, 1-L receiving flask, 1-L pear-shaped flask, feed tube with stopcock, vacuum seal, conical joint clip, and spherical joint clip.
Glass set 01
Vertical coil condenser, vertical coil adaptor, vapour tube for vertical condenser, 1-L receiving flask, 1-L pear-shaped flask, feed tube with stopcock, vacuum seal, conical joint clip, spherical joint clip, cooler holder, support bar clamp, and cooler support bar.
Glass set 02
Dewar condenser, vertical coil adaptor, vapour tube for vertical condenser, 1-L receiving flask, 1-L pear-shaped flask, feed tube with stopcock, vacuum seal, conical joint clip, spherical joint clip, cooler holder, support bar clamp, and cooler support bar.
Glass set 00P
Plastic-coated diagonal coil condenser, vapour tube for diagonal condenser, plastic-coated 1-L receiving flask, plastic-coated 1-L pear-shaped flask, feed tube with stopcock, vacuum seal, conical joint clip, and spherical joint clip.
Glass set 01P
Plastic-coated vertical coil condenser, plastic-coated vertical coil adaptor, vapour tube for vertical condenser, plastic-coated 1-L receiving flask, plastic-coated 1-L pear-shaped flask, feed tube with stopcock, vacuum seal, conical joint clip, spherical joint clip, cooler holder, support bar clamp, and cooler support bar.
Glass set 02P
Plastic-coated Dewar condenser, plastic-coated vertical coil adaptor, vapour tube for vertical condenser, plastic-coated 1-L receiving flask, plastic-coated 1-L pear-shaped flask, feed tube with stopcock, vacuum seal, conical joint clip, spherical joint clip, cooler holder, support bar clamp, and cooler support bar.
399,000.00 - ₹754,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Condenser Type Evaporation Flask Size Description
Availability Pricing
COMPARE
Diagonal 1 L Stuart® Rotary Evaporator w/ Diagonal Coil Condenser 1 L, 230 V 90C
₹399,000.00INR / Each
COMPARE
Vertical 1 L Stuart® Rotary Evaporator w/ Vertical Coil Condenser 1 L, 230 V 90C
₹436,000.00INR / Each
COMPARE
Cold Finger 1 L Stuart® Rotary Evaporator w/ Cold Finger Condenser 1 L, 230 V 90C
₹630,000.00INR / Each
COMPARE
Diagonal, Plastic Coated glassware 1 L Stuart® Rotary Evaporator w/ Diagonal Coil Condenser, Plastic Coated 1 L, 230 V 90C
₹630,000.00INR / Each
COMPARE
Vertical, Plastic Coated glassware 1 L Stuart® Rotary Evaporator w/ Vertical Coil Condenser, Plastic Coated 1 L, 230 V 90C
₹673,000.00INR / Each
COMPARE
Cold Finger, Plastic Coated glassware 1 L Stuart® Rotary Evaporator w/ Cold Finger Condenser, Plastic Coated 1 L, 230 V 90C
₹754,000.00INR / Each