Cole-Parmer® PVDF Tubing
Chemical- and abrasion-resistant tubing at a low cost
  • Resilient with good flexibility
  • Does not contain BPA, phthalates, or PVC
  • Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants
  • USP Class VI compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
With low extractable levels, high mechanical strength, and excellent abrasion and chemical resistance, PVDF tubing is an ideal choice for many critical fluid handling applications. Use this USP Class VI compliant, nonhemolytic tubing in pharmaceutical, food processing, and purified water applications. Tubing does not contain BPA, phthalates, or PVC that can leach out into the fluid, minimizing the risk of fluid contamination.
7,290.00 - ₹16,310.00INR / Pkg of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 1/8 1/32
₹7,290.00INR / Pkg of 1
COMPARE
1/8 3/16 1/32
₹10,850.00INR / Pkg of 1
COMPARE
1/8 1/4 1/16
₹16,310.00INR / Pkg of 1
COMPARE
3/16 1/4 1/32
₹11,180.00INR / Pkg of 1
COMPARE
5/32 1/4 3/64
₹14,630.00INR / Pkg of 1