Cole-Parmer PVDF Chromatography Syringe Filters
Low protein binding and high flow rates - ideal for filtration of difficult-to-filter solutions
  • Bidirectional flow with female luer lock on one side and a male luer slip on the other
  • Withstand pressure up to 100 psi (7 bar)
  • Batch test certificate included with each pack
PVDF Syringe Filters are both low protein binding and possess high flow rates, and offer great chemical compatibility—making them ideal for solutions that are difficult to filter. Best suited for applications requiring sterilization and clarification of biological solutions, and preparing HPLC solutions and aqueous samples. Maximum use temperature is 60°C (140°F).
3,200.00 - ₹3,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
13 Polyvinylidene fluoride (PVDF) 0.2
₹3,200.00INR / Each
COMPARE
13 Polyvinylidene fluoride (PVDF) 0.45
₹3,200.00INR / Each
COMPARE
25 Polyvinylidene fluoride (PVDF) 0.2
₹3,800.00INR / Each
COMPARE
25 Polyvinylidene fluoride (PVDF) 0.45
₹3,800.00INR / Each
COMPARE
33 Polyvinylidene fluoride (PVDF) 0.2
₹3,600.00INR / Each
COMPARE
33 Polyvinylidene fluoride (PVDF) 0.45
₹3,600.00INR / Each