Cole-Parmer® PTFE Syringe Filters
Polytetrafluoroethylene (PTFE) membranes are unaffected by most organic solvents, concentrated acids and bases, propellants, and cryogenic fluids
  • Ideal for sterilization of gas and alcohols; cleaning acids, alkalies, and organic solvents; venting purposes.
  • Use with aggressive chemicals
Filters are Triton-free and have a bidirectional membrane support and luer lock (15/25 mm dia) or stepped barbs (50 mm dia) inlet/outlet for a secure fit to the syringe. Membrane type and pore size are printed on each filter for easy identification. All Cole-Parmer syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards. Sterile filters are sterilized by gamma radiation. They are certified nonpyrogenic, noncytotoxic, and are blister packed.
12,700.00 - ₹57,000.00INR / Box of 20
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
COMPARE
25 0.2 Yes
₹31,000.00INR / Box of 50
COMPARE
25 0.2 No
₹31,800.00INR / Box of 100
COMPARE
25 0.45 No
₹39,000.00INR / Box of 100
COMPARE
50 0.2 No
₹16,700.00INR / Box of 20
COMPARE
50 0.45 No
₹12,700.00INR / Box of 20
COMPARE
15 0.2 No
₹57,000.00INR / Pkg of 100
COMPARE
15 0.45 No
₹26,400.00INR / Pkg of 100
Showing 1- 7 of 7