Cole-Parmer® Pressure/Vacuum System Components
Customize to your needs
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
Sensors provide improved tracking ability and maintain calibration over long periods of time. When replacing a sensor, you do not need to recalibrate.
These sensors are suited for use with mercury and oil pumps.
Pirani type sensors offer enhanced stability and an automatic temperature compensation network. The 304 stainless steel sensors operate at half the
temperature of standard thermopile sensors. Use for high-vacuum applications.
Diaphragm type sensor offers quick response—less than 400 milliseconds full-scale. Select a diaphragm type sensor made from 316 stainless steel or silicon based on your application.
35,500.00 - ₹102,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accuracy Resolution Description
Availability Pricing
COMPARE
±5% of reading or ±0.03 mTorr 0.01 mTorr 0.01 To 100 Mtorr Pressure/vacuum Pirani-type Sensor, Nw 16 Connection
Call For Price
COMPARE
±5% of reading or ±3 mTorr 1 mTorr 1 To 2000 Mtorr Pressure/vacuum Pirani-type Sensor, Nw 16 Connection
₹45,500.00INR / Each
COMPARE
±5% of reading or ±0.03 mTorr 0.01 mTorr 0.01 To 100 Mtorr Pressure/vacuum Pirani-type Sensor, 3/4" Tube OD
₹53,300.00INR / Each
COMPARE
±5% of reading or ±3 mTorr 1 mTorr 1 To 2000 Mtorr Pressure/vacuum Pirani-type Sensor, 1/8" NPT(M)
₹35,500.00INR / Each
COMPARE
±5% of reading or ±0.03 Torr 0.01 Torr 0.01 To 20 Torr Pressure/vacuum Pirani-type Sensor, 1/8" NPT(M)
₹47,500.00INR / Each
COMPARE
±5% of reading or ±3 mTorr 1 mTorr 1 to 2000 mtorr Pressure/Vacuum Calibrator for Pirani-Type Sensor
₹102,000.00INR / Each