Cole-Parmer 0.01 To 20 Torr Pressure/vacuum Pirani-Type Sensor, 1/8" NPT(M)
Representative image only.
Cole-Parmer 0.01 To 20 Torr Pressure/vacuum Pirani-Type Sensor, 1/8" NPT(M)
Cole-Parmer | Item: 6880152
Customize to your needs
₹47,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹47,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer 0.01 To 20 Torr Pressure/vacuum Pirani-type Sensor, 1/8" NPT(M)
Cole-Parmer 0.01 To 20 Torr Pressure/vacuum Pirani-type Sensor, 1/8" NPT(M)
6880152
₹47,500.00INR / Each
+ + +
Cole-Parmer Vacuum Sensor Extension Cable, 20'
6880175
₹31,400.00INR / Each
=
Total Price 244600.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
More About this Item
Sensors provide improved tracking ability and maintain calibration over long periods of time. When replacing a sensor, you do not need to recalibrate.
These sensors are suited for use with mercury and oil pumps.
Pirani type sensors offer enhanced stability and an automatic temperature compensation network. The 304 stainless steel sensors operate at half the
temperature of standard thermopile sensors. Use for high-vacuum applications.
Diaphragm type sensor offers quick response—less than 400 milliseconds full-scale. Select a diaphragm type sensor made from 316 stainless steel or silicon based on your application.
Specifications
  • Accuracy±5% of reading or ±0.03 Torr
  • Wetted Materials304 SS
  • Process Connection1/8" NPT(M)
  • Media CompatibilityGases compatible with wetted materials
  • Resolution0.01 Torr
  • Description0.01 To 20 Torr Pressure/vacuum Pirani-type Sensor, 1/8" NPT(M)