Cole-Parmer®Presaturated IPA Surface Tub Wipes
Durable, tear-resistant, lint-free polyester and viscose Presaturated wipes
  • 70% polyester, 30% viscose
  • High abrasion resistance
  • Consistent saturation of 70% IPA and 30% DI water
  • Convenient flip lid canister with pull-through dispense
  • Non-sterile; packaged in an ISO Class 8 cleanroom
Cole-Parmer® Presaturated disposable wipes are composed of 70% polyester and 30% viscose, pre-saturated in a solution of 70% isopropyl alcohol (IPA) and 30% deionized (DI) water. Ideal for the quick removal of surface-level contaminants from workbenches, electronics, equipment and tools. Suitable for a variety of controlled environments, including ISO Class 8 cleanrooms.
14,300.00 - ₹17,000.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Presaturated Wipes, 70% IPA 30% DI, 7\"X8\", 45 gsm, non-sterile; 100/Canister
₹14,300.00INR / Pkg of 100
COMPARE
Presaturated Wipes, 70% IPA 30% DI, 8\"X8\", 45 gsm, non-sterile; 100/Canister
₹17,000.00INR / Pkg of 100