Cole-Parmer® Precleaned EPA Vials
Precleaned to meet strict EPA requirements
  • Constructed of durable amber or clear glass
These glass vials are precleaned to meet strict EPA requirements. Vials with certificate of analysis meet EPA criteria for more than 60 volatile organic analyses (VOA). All vials include a polypropylene cap with a 1/8 inch PTFE-faced silicone septum.
17,900.00 - ₹41,000.00INR / Case of 72
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Septum Type Tube Color Volume (mL)
Availability Pricing
COMPARE
PTFE/silicone Clear 20
₹17,900.00INR / Case of 72
COMPARE
PTFE/silicone Clear 20
₹18,400.00INR / Case of 72
COMPARE
PTFE/silicone Clear 40
₹41,000.00INR / Case of 72
COMPARE
PTFE/silicone Amber 40
₹26,300.00INR / Case of 72
COMPARE
PTFE/silicone Clear 40
₹32,600.00INR / Case of 72
COMPARE
PTFE/silicone Amber 40
₹38,400.00INR / Case of 72
COMPARE
PTFE/silicone Clear 60
₹22,400.00INR / Case of 72
COMPARE
PTFE/silicone Amber 60
₹28,500.00INR / Case of 72
COMPARE
PTFE/silicone Clear 60
₹27,400.00INR / Case of 72
COMPARE
PTFE/silicone Amber 60
₹31,500.00INR / Case of 72
Showing 1- 10 of 10