Cole-Parmer® Porcelain Round-Form Evaporating Dishes
Perfect for samples with high liquid content
  • No softening occurs up to 1000 °C (1832 °F)
  • Resistance to chemical action
  • Thermal shock resistance
  • Autoclavable
Round-form evaporating dishes feature lip and spout. They can withstand temperatures to 1150 °C (2102 °F) and are resistant to acids and alkalies except for hydrofluoric acid. Dishes up to 250 mL capacity are glazed inside and out. Dishes 385 mL and greater are glazed inside and partially glazed outside.
4,500.00 - ₹138,000.00INR / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Height (in) Top Outside Diameter (in)
Availability Pricing
COMPARE
35 15/16 2 3/8
₹7,100.00INR / Pkg of 6
COMPARE
70 1 3/16 2 15/16
₹7,200.00INR / Pkg of 6
COMPARE
80 1 3/16 3 1/8
₹7,600.00INR / Pkg of 6
COMPARE
120 1 5/8 3 3/4
₹9,800.00INR / Pkg of 6
COMPARE
150 1 5/8 3 15/16
₹14,000.00INR / Pkg of 6
COMPARE
250 1 3/4 4 9/16
₹14,000.00INR / Pkg of 6
COMPARE
385 1 7/8 5 3/4
₹4,500.00INR / Each
COMPARE
525 2 6 3/8
₹6,600.00INR / Each
COMPARE
765 2 1/8 7 1/4
₹9,100.00INR / Each
COMPARE
1285 2 15/16 8 1/2
₹13,100.00INR / Each
COMPARE
2100 3 1/8 10 7/16
₹17,300.00INR / Each
COMPARE
3250 3 3/4 12
₹19,300.00INR / Each
COMPARE
35 15/16 2 3/8
₹50,100.00INR / Pkg of 12
COMPARE
35 15/16 2 3/8
₹138,000.00INR / Pkg of 48
COMPARE
80 1 3/16 3 1/8
₹50,700.00INR / Pkg of 24
COMPARE
120 1 5/8 3 3/4
₹80,100.00INR / Pkg of 24
COMPARE
150 1 5/8 3 15/16
₹60,300.00INR / Pkg of 18
COMPARE
250 1 3/4 4 9/16
₹69,100.00INR / Pkg of 18
COMPARE
525 2 6 3/8
₹102,000.00INR / Pkg of 12
COMPARE
1285 2 15/16 8 1/2
₹83,500.00INR / Pkg of 6
Showing 1- 20 of 20