Cole-Parmer® Porcelain Flat-Form Evaporating Dishes
Shallow bowl dries solid samples effectively
  • Autoclavable
Flat-form dishes are glazed inside and out, except the rim and part of the outside bottom, and they feature a lip and spout.
2,900.00 - ₹82,500.00INR / Pkg of 6
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Height (in) Top Outside Diameter (in)
Availability Pricing
COMPARE
35 5/8 2 1/2
₹9,100.00INR / Pkg of 6
COMPARE
50 3/4 2 15/16
₹9,900.00INR / Pkg of 6
COMPARE
80 7/8 3 3/8
₹11,300.00INR / Pkg of 6
COMPARE
100 55/64 4 21/64
₹15,000.00INR / Pkg of 6
COMPARE
200 1 3/8 4 3/4
₹17,400.00INR / Pkg of 6
COMPARE
300 1 1/4 5 15/16
₹36,200.00INR / Pkg of 6
COMPARE
100 15/16 3 15/16
₹2,900.00INR / Each
COMPARE
200 1 3/8 4 3/4
₹5,900.00INR / Each
COMPARE
290 1 1/4 5 15/16
₹11,000.00INR / Each
COMPARE
35 5/8 2 1/2
₹66,000.00INR / Pkg of 24
COMPARE
50 3/4 2 15/16
₹31,300.00INR / Pkg of 24
COMPARE
100 15/16 3 15/16
₹82,500.00INR / Pkg of 18
COMPARE
200 1 3/8 4 3/4
₹63,500.00INR / Pkg of 12
Showing 1- 13 of 13