Cole-Parmer® Porcelain Deep-Form Mortar and Pestle Sets
No more sample loss – high sides and narrow opening contain particulates
  • Autoclavable
Mortar features lip and spout, and is glazed on the outside (except the bottom). Oversized pestle is also glazed (except the grinding surface). Mortar and pestle are sold as a set.
2,200.00 - ₹90,600.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Mortars Capacity (mL) Mortars Height (mm) Mortars Top OD (mm)
Availability Pricing
COMPARE
150 70 90
₹42,500.00INR / Pkg of 12
COMPARE
275 70 115
₹34,800.00INR / Pkg of 8
COMPARE
400 80 130
₹46,800.00INR / Pkg of 8
COMPARE
1900 130 210
₹90,600.00INR / Pkg of 2
COMPARE
50 47 70
₹2,200.00INR / Each
COMPARE
70 52 80
₹2,500.00INR / Each
COMPARE
150 70 90
₹5,300.00INR / Each
COMPARE
275 70 115
₹4,100.00INR / Each
COMPARE
400 80 130
₹6,000.00INR / Each
COMPARE
750 110 163
₹7,900.00INR / Each
COMPARE
1900 130 210
₹17,200.00INR / Each
Showing 1- 11 of 11