Cole-Parmer® Polysulfone Tri-Clamp Sight Gauges
Visually confirm flow, color, and clarity of your process media
  • PSU offers superior optical clarity
  • Sanitary quick clamp connections
  • Sterilize by gamma irradiation or steam
  • ASME-BPE, USP Class VI, ISO 10993 (Sections 4 and 5), FDA 21 CFR 177.1655, REACH, and RoHS compliant
  • Animal-derived ingredient free (ADIF) and animal-derived component free (ADCF)
Get an unobstructed view of your sample on a large, clear polysulfone sight gauge. The large tubular window offers a view from multiple angles. The sight gauges withstand temperatures ranging from -29 to 155°C (-20 to 240°F) and exhibit excellent chemical resistance, making them perfect for critical bioprocessing, food processing, and chemical processing applications. Simply use sanitary Tri-Clamp fittings and clamps for leakproof, sterile connections.
112,000.00 - ₹200,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
End Fitting Length (InchShort) Tubing ID (in)
Availability Pricing
COMPARE
3/4" Sanitary Connection 6 31/64
₹112,000.00INR / Each
COMPARE
1" Sanitary Connection 6 55/64
₹156,000.00INR / Each
COMPARE
1-1/2" Sanitary Connection 6 1 23/64
₹164,000.00INR / Each
COMPARE
2" Sanitary Connection 6 1 55/64
₹178,000.00INR / Each
COMPARE
2-1/2" Sanitary Connection 6 2 23/64
₹190,000.00INR / Each
COMPARE
3" Sanitary Connection 6 2 55/64
₹200,000.00INR / Each