Cole-Parmer Sight Gauge, Polysulfone, 6"L, 3/4" Sanitary Connection
Representative image only.
Cole-Parmer Sight Gauge, Polysulfone, 6"L, 3/4" Sanitary Connection
Cole-Parmer | Item: 5014283
Visually confirm flow, color, and clarity of your process media
₹112,000.00INR / Each
₹112,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Sight Gauge, Polysulfone, 6"L, 3/4" Sanitary Connection
Cole-Parmer Sight Gauge, Polysulfone, 6"L, 3/4" Sanitary Connection
5014283
₹112,000.00INR / Each
+ =
Total Price 114700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • PSU offers superior optical clarity
  • Sanitary quick clamp connections
  • Sterilize by gamma irradiation or steam
  • ASME-BPE, USP Class VI, ISO 10993 (Sections 4 and 5), FDA 21 CFR 177.1655, REACH, and RoHS compliant
  • Animal-derived ingredient free (ADIF) and animal-derived component free (ADCF)
  • 3/4" sanitary connection
More About this Item
Get an unobstructed view of your sample on a large, clear polysulfone sight gauge. The large tubular window offers a view from multiple angles. The sight gauges withstand temperatures ranging from -29 to 155°C (-20 to 240°F) and exhibit excellent chemical resistance, making them perfect for critical bioprocessing, food processing, and chemical processing applications. Simply use sanitary Tri-Clamp fittings and clamps for leakproof, sterile connections.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Tubing FormulationPolysulfone
  • Tubing ID (in)31/64
  • Tubing ID (mm)12.3
  • End Fitting3/4" Sanitary Connection
  • Sterilization MethodAutoclave;Gamma;EtO
  • Regulatory ApprovalsADCF;FDA;USP;RoHS
  • Length (InchShort)6
  • DescriptionSight Gauge, Polysulfone, 6"L, 3/4" Sanitary Connection