Cole-Parmer® Polypropylene Filter Holders
Reusable filter holders with no tear design save time and money
  • Constructed of durable polypropylene for enhanced pressure and temperature applications
  • Efficiently assemble holders without membrane tears
  • Ideal for ultra-cleaning and sterilizing liquids under pressure, or for aseptic sampling of liquids and gases
  • Autoclavable
These polypropylene (PP) filter holders assemble quickly and efficiently with a specially designed inlet cap and exterior lock. This allows for ease of use without damaging or tearing membranes.
Suitable for use with liquids and gases these holders feature a maximum operating temperature (liquids) of 176°F (80°C). The 25-mm holder has a maximum operating pressure of 42 psi (2.9 bar) and 71 psi (4.8 bar) for the 47-mm holder. Both are autoclavable.
6,500.00 - ₹13,000.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Inlet Connection Outlet Connections
Availability Pricing
COMPARE
47 Female luer-slip Male luer-slip
₹13,000.00INR / Pkg of 10
COMPARE
25 Female luer slip Male luer slip
₹6,500.00INR / Pkg of 10