Cole-Parmer®Polyester Presaturated Cleanroom Wipes
Sterile Polycellulose wipes Presaturated with 70% Isopropyl Alcohol
  • Durable 100% continuous filament, double-knit polyester fiber
  • Premoistened with 70% isopropyl alcohol and 30% DI/WFI
  • 120 GSM, Laser cut edge for reduced fiber contamination
  • Very low levels of particulate and extractable counts
  • High abrasion resistance
  • Gamma sterilized and packaged in an ISO Class 5 cleanroom
  • Convenient resealable pouch
Formats:
• Pack sizes - Pack of 10 or Pack of 15 or Pack of 20
• Wipes Size - Available in 9’’ x 9’’ and 12’’ x 12’’
• Water Component – Available in 30% DI water or 30% WFI water
Cole-Parmer® Presaturated wipes contain 100% continuous filament polyester. Presaturated in a solution of 70% isopropyl alcohol (IPA) and either 30% deionized (DI) water or 30% water for injection (WFI).
Ideal for the quick removal of surface-level contaminants from workbenches, electronics, equipment and tools. Suitable for a variety of controlled environments, including ISO Class 5 cleanrooms.
46,400.00 - ₹81,000.00INR / Case of 400
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Polyester IPA Wipes 70%IPA 30% DI; 9"x9", 120 gsm, Gamma Sterile; 10/Pk, 40 Pack/Cs
₹51,000.00INR / Case of 400
COMPARE
Polyester IPA Wipes, 70% IPA 30% DI, 12"x12", 120 gsm, Gamma Sterile; 15/Pk, 40 Pack/Cs
₹81,000.00INR / Case of 600
COMPARE
Polyester IPA Wipes, 70%IPA 30% DI, 12"x12", 120 gsm, Gamma Sterile; 10/Pk, 40 Pack/Cs
₹54,200.00INR / Case of 400
COMPARE
Polyester IPA Wipes, 70%IPA 30% WFI, 9"x 9", 120 gsm, Gamma Sterile; 15/Pk, 26 Pack/Cs
₹46,400.00INR / Case of 390
COMPARE
Polyester IPA Wipes, 70%IPA 30% WFI; 9"x 9", 120 gsm, Gamma Sterile; 10/Pk, 40 Pack/Cs
₹51,000.00INR / Case of 400
COMPARE
Polyester IPA Wipes, 70%IPA 30% WFI; 12"x12", 120 gsm, Gamma Sterile; 20/Pk, 20 Pack/Cs
₹52,700.00INR / Case of 400
COMPARE
Polyester IPA Wipes, 70%IPA 30% WFI; 12"x12", 120 gsm, Gamma Sterile; 10/Pk, 40 Pack/Cs
₹54,200.00INR / Case of 400
Showing 1- 7 of 7