Cole-Parmer® Polyester Cleanroom Wipes
Laundered Polyester Knits deliver the low particles, fibres and NVRs
  • Woven 100% polyester fibre with laser sealed edge for minimal lint and particle release
  • Low non-volatile residue (NVR), fibres, ions and extractables
  • Weight: 140 ± 5 GSM
  • Excellent chemical resistance and compatible with a variety of solutions
  • Abrasion resistant - won’t tear under rigorous use
  • Highly absorbent with critically low levels of particulate counts
  • Lot to lot traceable
  • Laundered and packaged in a cleanroom class 100
Cole-Parmer® Polyester Cleanroom Wipes are manufactured from high strength, continuous filament 100% polyester. Polyester cleanroom wipes are soft, durable, and highly absorbent making quick, effective work of wiping and cleaning surfaces in your critical environment.
Choose from sterile gamma irradiated and non-sterile versions.. Available sizes include 9’’ x 9’’ (228 x 228 mm) and 12’’ x 12’’ (304 x 304 mm).
Application:
• Wiping and cleaning surfaces, equipment and parts
• Applying and removing lubricants, adhesives, disinfectants and other solutions and solutions including disinfectants
• Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
• Cleaning or disinfecting workstation surfaces and processing equipment
10,700.00 - ₹15,400.00INR / Case of 1500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Cleanroom Wipes, Polyester, 9" x 9", Non-Sterile; 150 pcs/Bag, 10 Bags/Case
₹10,700.00INR / Case of 1500
COMPARE
Cleanroom Wipes, Polyester, 9" x 9", Sterile; 150 pcs/Bag, 10 Bags/Case
₹11,600.00INR / Case of 1500
COMPARE
Cleanroom Wipes, Polyester, 12" x 12", Non-Sterile; 150 pcs/Bag, 10 Bags/Case
₹14,500.00INR / Case of 1500
COMPARE
Cleanroom Wipes, Polyester, 12" x 12", Sterile; 150 pcs/Bag, 10 Bags/Case
₹15,400.00INR / Case of 1500