Cole-Parmer Poly-Cellulose Wipes
Offer exceptional durability and chemical resistance as well as high absorbency
  • Made of 45% polyester/55% cellulose composite
  • Weight: 68 g/m²
  • Highly absorbent
  • Low levels of particles and and extractables
  • Lot-to-lot traceable
Cole-Parmer® poly-cellulose wipes are ideal for general cleanroom wiping and spill control. These non-woven wipes combine the features of exceptional durability, chemical resistance achieved by the low linting characteristics of polyester with the high liquid retention rates from the properties of cellulose.
Choose from sterile gamma irradiated and non-sterile versions.. Available sizes include 9’’ x 9’’ (228 x 228 mm) and 12’’ x 12’’ (304 x 304 mm).
Applications:
• Ideal for wiping scratch sensitive surfaces in cleanrooms
• Excellent for cleaning and polishing of delicate equipment
• Cleaning or disinfecting workstation surfaces and processing equipment
• Applying or removing cleaning and disinfecting solutions
• Cleaning optical machinery, flat panel displays and tools
10,600.00 - ₹28,000.00INR / Case of 3000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Description
Availability Pricing
COMPARE
45% Polyester / 55% Cellulose composite, Non Sterile Cole-Parmer Poly-Cellulose Wipes, 9\" x 9\", Non-Sterile, 68 GSM; 300/Bag, 10 Bags/Case
₹10,600.00INR / Case of 3000
COMPARE
45% Polyester / 55% Cellulose composite, Sterile Cole-Parmer Poly-Cellulose Wipes, 9\" x 9\", Sterile, 68 GSM; 300/Bag, 10 Bags/Case
₹28,000.00INR / Case of 3000
COMPARE
45% Polyester / 55% Cellulose composite, Non Sterile Cole-Parmer Poly-Cellulose Wipes, 12\" x 12\", Non-Sterile, 68 GSM; 300/Bag, 10 Bags/Case
₹19,300.00INR / Case of 3000
COMPARE
45% Polyester / 55% Cellulose composite, Sterile Cole-Parmer Poly-Cellulose Wipes, 12\" x 12\", Sterile, 68 GSM; 300/Bag, 10 Bags/Case
₹23,200.00INR / Case of 3000