Cole-Parmer® Essentials PETG Square Media Bottles
Reduce risk of cross contamination with disposable Cole-Parmer media bottles
  • Softer, shock resistant and pliable PETG material ensures comfortable grip
  • Supplied with HDPE (High Density Polyethylene) screw caps—free of heavy metal
  • Bottle is embossed with graduations on both the sides
  • 3-years shelf life from the month of production
  • All used materials meet the legal requirements of FDA
  • Temperature range from -60°C to 60°C
  • Free of detectable DNase/RNase, human DNA and pyrogens
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Capacity (mL) Sterile
Availability Pricing
COMPARE
500 500 No
Call For Price
COMPARE
500 500 Yes