Cole-Parmer® Peroxide-Cured Silicone Tubing
Flexible tubing offers long shelf life and resists kinking and tearing
  • Exhibits good heat, cold, ozone, UV, and abrasion resistance
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Peroxide-cured silicone tubing provides a long shelf life and enhanced mechanical strength—excellent for use in peristatic pumps. Featuring a smooth bore, the tubing offers low coefficient of friction for effortless fluid transfer.
4,930.00 - ₹13,350.00INR / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 3/16 1/16
₹4,930.00INR / Pkg of 1
COMPARE
1/8 1/4 1/16
₹5,450.00INR / Pkg of 1
COMPARE
3/16 5/16 1/16
₹6,090.00INR / Pkg of 1
COMPARE
1/4 3/8 1/16
₹6,740.00INR / Pkg of 1
COMPARE
3/8 1/2 1/16
₹7,780.00INR / Pkg of 1
COMPARE
3/16 7/16 1/8
₹9,720.00INR / Pkg of 1
COMPARE
1/4 1/2 1/8
₹10,760.00INR / Pkg of 1
COMPARE
3/8 5/8 1/8
₹13,350.00INR / Pkg of 1
Showing 1- 8 of 8