Cole-Parmer® Orbital Sieve Shakers
Back-and-forth lateral motion combined with up/down and tilting actions distribute material across the sieve
  • Accurately separate particles from size no. 4 to no. 200 (4.75 mm to 75 µm)
  • Precisely time operation up to 99 minutes with the built-in digital countdown timer
  • EZ-Clamp system quickly and securely locks in sieves for testing
  • Compliant with multiple ASTM and AASHTO standards
Use the orbital sieve shakers for particle size testing of mineral aggregates and for sieve gradation of free-flowing granular materials. The back-and-forth and up/down motions plus tilting action move the testing samples across the entire mesh area of the test sieve to quickly separate the materials. Orbital shakers accommodate from no. 4 to no. 200 material size range but may be used with larger sizes when specifications permit.
Shakers feature a digital timer with a large LED display that can be seen from a distance. Use the countdown timer for operations up to 99 minutes with 1 second increment. Timer includes a pause function and user-selectable modes for great versatility. Easy one-button operation and saved settings offer excellent repeatability between cycles.
Quickly and easily lock in the sieves using the EZ-Clamp system. The sieve cover slides freely up and down the clamp rods to hold sieves in place. A quick twist of the clamping screws secures the stack in place. All mechanical and electrical parts are enclosed in a rugged painted steel case for safety and durability.
393,000.00 - ₹474,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Sieves Size Accepted (OD)
Availability Pricing
COMPARE
8" or 200 mm
₹393,000.00INR / Each
COMPARE
8", 12", 200 mm, or 300 mm
₹474,000.00INR / Each