Cole-Parmer® Narrow-Mouth Plastic Bottles, HDPE
Chemical-resistant — stands up to most corrosives
  • Withstands freezer storage down to -100°C
  • Will not leak with PP screw caps
  • Meets USP Class VI requirements
These all-purpose bottles are popularly used for packaging, shipping, and storage because of their reliable durability. Rigid, translucent, and chemical-resistant, they can be used with most corrosives. All have leakproof polypropylene screw caps.
2,800.00 - ₹8,400.00INR / Pkg of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 20 mm High Density Polyethyene (HDPE)
₹2,800.00INR / Pkg of 12
COMPARE
60 20 mm High Density Polyethyene (HDPE)
₹3,200.00INR / Pkg of 12
COMPARE
125 24 mm High Density Polyethyene (HDPE)
₹3,700.00INR / Pkg of 12
COMPARE
250 24 mm High Density Polyethyene (HDPE)
₹5,400.00INR / Pkg of 12
COMPARE
500 24 mm High Density Polyethyene (HDPE)
₹8,400.00INR / Pkg of 12
COMPARE
1000 38 mm High Density Polyethyene (HDPE)
₹6,800.00INR / Pkg of 6